missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SP 141
GREENER_CHOICE

Product Code. p-5545605
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778108 10 mg 10mg
18716539 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18778108 Supplier Tocris Bioscience™ Supplier No. 5332/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity MDM2 inhibitor

SP 141 is a high affinity MDM2 inhibitor (Ki = 28 nM); directly binds MDM2. SP 141 inhibits MDM2 expression and induces MDM2 autoubiquitination and proteasomal degradation. SP 141 attenuates growth of breast cancer xenograft tumors. SP 141 impairs liposarcoma growth in vitro and in human xenograft models. SP 141 is cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SP 141
Purity >98%
Molecular Weight (g/mol) 324.38

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.