missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SP 600125

Product Code. 30000032
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000032 Supplier Tocris Bioscience™ Supplier No. TB1496RMU/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SP 600125 synthesized to Ancillary Material Grade

TB1496-RMU is an Ancillary Materials Grade version of SP 600125 (Cat. No. 1496). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Find out more about our Ancillary Material Grade product range.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anthra[1,9-cd]pyrazol-6(2H)-one
CAS 129-56-6
Quantity 10 mg
Target JNK/c-jun Inhibitors
Molecular Formula C14H8N2O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.