missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SP 600125
GREENER_CHOICE

Product Code. 18095862
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095862 10 mg 10mg
18095572 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095862 Supplier Tocris Bioscience™ Supplier No. 1496/10
  • ONLINE EXCLUSIVE PRICE
    170.00 €
    160.65 € / 10mg
    Save 9.35 €
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective JNK inhibitor

SP 600125 is a selective JNK inhibitor. Competitively and reversibly inhibits JNK1, 2 and 3 (IC50 = 40 - 90 nM) with negligible activity at ERK2, p38β and a range of enzymes. Active in vivo. Exhibits reduced selectivity over other protein kinases under certain conditions. SP 600125 protects renal tubular epithelial cells from ischemia/reperfusion-induced apoptosis. Promotes chondrogenesis of bone mesenchymal stem cells by inhibition of TNF-α-induced inflammation. Adipogenesis from MSCs is also promoted by SP 600125. Essential component of medium for maintaining stem cells in naive pluripotent state.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-56-6
Molecular Formula C14H8N2O

Specifications

Quantity 10 mg
Formula Weight 220.23
Purity >98%
Molecular Weight (g/mol) 220.231
Chemical Name or Material SP 600125
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.