missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Shop All SP Scienceware Products
Click to view available options
Color:
Clear
Clear with Blue Ends
Unit Size:
Each
Description
- Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Stack up to 3 units high to save space
- Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Cabinet Style | Vertical |
| Material | Copolyester |
| Product Type | Desiccator |
| Door Style | Left-hinged and gasketed |
| For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 |
| Includes | Carrying handles, one latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer |
| Model | 1.0 |
| No. of Shelves | 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction