missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SP Sepharose™ XL

Produktkod. 11530814 Handla allt Cytiva Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
300 mL
Förpackningsstorlek:
300mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11530814 300 mL 300mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11530814 Leverantör Cytiva Leverantörsnummer 17507301

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Strong cation exchanger with high loading capacity and chemical stability

  • Bioprocess medium with hydrophilic base matrix
  • Ligand: sulphopropyl
  • Average particle size: 90μm
  • Matrix: 6% cross-linked agarose with dextran surface extenders
  • Ionic capacity: 0.18 to 0.25mmol H+/mL medium
  • pH stability working range: 4 to 13
  • For use in capture steps of biomolecules from clarified feedstocks
  • Available in 300mL bottle

TRUSTED_SUSTAINABILITY

Specifikationer

Quantity 300 mL
Stability Non-ionic detergents, 1 M NaOH, 6 M Guanidine Hydrochloride
For Use With (Application) For Strong CatIon Exchanger with High Loading Capacity and Chemical Stability
Storage Requirements 4°C to 30°C
Product Type SP Sepharose XL Media
Flow Rate 300 to 500 cm/Hoμr
Particle Size 45 to 165 μm
Ionic Capacity 0.18 to 0.25 mM H+/mL
Includes Certificate of Analysis
Ion Exchange Type Strong CatIon Exchanger
Matrix 6% Cross-Linked Agarose with Dextran surface extenders
pH Range 4 to 13
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphopropyl
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • SP Sepharose XL, 300 ml
Signalord
  • Warning
Riskkategorier
  • Flammable liquid Category 3
Faroangivelser
  • H226-Flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Kompletterande information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.