Learn More
Agilent BioTek Spare AMX manifold
Spare AMX manifold set, contains aspirate and dispense manifolds
Brand: Agilent BioTek 1260036
Additional Details : Weight : 1.00000kg
Description
- Includes one dispense and one aspirate manifold
- For use with 1260035 (one each), sold separately
Specifications
CE and TUV marked. RoHS compliant. | |
Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization | |
Spare AMX manifold set, aspirate and dispense manifolds |
Spare AMX manifold set | |
MFXP2R |