Learn More
Brady™ SPC™ Absorbents Acid Neutralizing Chemical Specialty Spill Kit
Product Code. 30533401 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
4.491 gal.
Unit Size:
1 set
Product Code. Capacity (English) Unit Size
30533401 4.491 gal. 1 set
1 options
Product Code. 30533401 Supplier Brady™ Supplier No. 241138
  • ONLINE EXCLUSIVE PRICE
    220.00 €
    191.12 € / 1 sets
    Save 28.88 €
    13% Off
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Position spill kits around your facility so that you are prepared should a spill occur. These kits combine pads, PPE and accessories so that you are prepared in an emergency.
Position spill kits around your facility so that you are prepared should a spill occur.
  • These kits combine pads, PPE and accessories so that you are prepared in an emergency.
  • This spill kit contains chemical neutralizing polymers which provide safe spill control with a color change indicator that reflects neutralisation.
  • Provides safe spill control for most acids (including battery spills on smooth surfaces).
Specifications
Capacity (English) 4.491 gal.
Type Spill Kit
Includes 2 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper
Model Chemical Sorbent
Alternate Part Number SKA-BKTACID-E