missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents ALLWICK Universal Maintenance ADR-L Spill Kit

Product Code. 30533307
Change view
Click to view available options
Capacity (English):
14265 gal.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) unitSize
30533307 14265 gal. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533307 Supplier Brady™ Supplier No. 813866
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

SKA-ADR-L is a portable spill kit that comes in a durable see-through bag with shoulder strap. Designed for small spills of industrial liquids including oils, water, solvents, coolants, etc. Requires a minimum of storage space yet it contains all the sorbents needed to handle a small spill.

SKA-ADR-L is a portable spill kit that comes in a durable see-through bag with shoulder strap.

  • Designed for small spills of industrial liquids including oils, water, solvents, coolants, etc.
  • Requires a minimum of storage space yet it contains all the sorbents needed to handle a small spill: socs to contain the spill - pads and pillows for the clean up.
  • Durable see-through bag makes it easy to check the spill response supplies.
  • Quick and easy access to the sorbents ensuring a fast response thanks to the zipper which opens up the entire front of the bag.
  • Contents: 35 pads (41 x 51 cm) , 1 soc (7.6 cm x 122 cm), 1 soc (7,6 cm x 244 cm), 1 pillow (43 x 48 cm), 4 pillows (25 x 25 cm), 1 pair of gloves and 2 disposal bags.

Specifications

Color Yellow
Capacity (English) 14265 gal.
Type Spill Kit
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Process, Warehouse and Logistics
For Use With (Equipment) Emergency Spill Response
Includes 35 Pads (40 x 50 cm), 1 Socs (7.6 cm dia. x 122 cm), 1 Soc (7.6 dia. cm x 244 cm), 1 Pillow (43 x 48 cm), 4 Pillows (23 x 23 cm), 1 Pair of Gloves, Instruction Sheet and 2 Disposal Bags
Model Universal Sorbent
Alternate Part Number SKA-ADR-L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.