Learn More
Brady™ SPC™ Absorbents Base Neutralizing Chemical Specialty Spill Kit
Product Code. 30533372 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
4.491 gal.
Unit Size:
1 set
Product Code. Capacity (English) Unit Size
30533372 4.491 gal. 1 set
1 options
Product Code. 30533372 Supplier Brady™ Supplier No. 241139
  • ONLINE EXCLUSIVE PRICE
    213.90 €
    198.76 € / 1 sets
    Save 15.14 €
    7% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Position Spill Kits around your facility so that you are prepared should a spill occur. These kits combine pads, accessories and PPE so that you are prepared in an emergency. This spill kit contains chemical neutralizing polymers which provide safe spill control with a color change indicator that reflects neutralization.
Position spill kits around your facility so that you are prepared should a spill occur.
  • These kits combine pads, accessories and PPE so that you are prepared in an emergency.
  • This spill kit contains chemical neutralizing polymers which provide safe spill control with a color change indicator that reflects neutralisation.
  • Use to neutralize highly basic substances before clean-up.
Specifications
Capacity (English) 4.491 gal.
Type Spill Kit
Includes 2 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x AW412 (813789), 10 x GP100-M (813812), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS, (198790) 1 x SPC-BASE, 1 x Bucket, 1 x Scoop and Scraper
Model Chemical Sorbent
Alternate Part Number SKA-BKTBASE-E