Learn More
Brady™ SPC™ Absorbents Battery Acid Specialty Spill Kit
Product Code. 30533339 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
5.283 gal.
Unit Size:
1 set
Product Code. Capacity (English) Unit Size
30533339 5.283 gal. 1 set
1 options
Product Code. 30533339 Supplier Brady™ Supplier No. 241135
  • ONLINE EXCLUSIVE PRICE
    189.20 €
    175.85 € / 1 sets
    Save 13.35 €
    7% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Position the Battery Acid Spill Kits near a battery swap station and around your facility so that you are prepared should a spill occur. This spill kit is designed to pick up battery acid on both smooth and rough surfaces while remaining chemically inert.
Position the battery acid spill kits near a battery swap station and around your facility so that you are prepared should a spill occur.
  • This spill kit is designed to pick up battery acid on both smooth and rough surfaces while remaining chemically inert.
  • These kits combine pads, spillfix, PPE and accessories so that you are prepared in an emergency.
  • This Spill kit contains chemical neutralizing polymers which provide safe spill control with a color change indicator that reflects neurtalisation.
Specifications
Capacity (English) 5.283 gal.
Type Spill Kit
Includes 2 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper, 1 x SF-JAR-CS (149265)
Model Acids
Alternate Part Number SKH-BATT-E