missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents BrightSorb High Visibility Safety Heavyweight Absorbent Pad

Product Code. 30533356
Change view
Click to view available options
Capacity (Metric):
81 L
155 L
263 L
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30533356 81 L Pack of 100
30533341 155 L Each
30533313 263 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30533356 Supplier Brady™ Supplier No. 309070
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

MAXX means greater performance without the extra weight. Specially developed chemical sorbent, yellow color-coded for safety and easy separation of hazardous waste, heavy weight pad. Does not react with aggressive fluids.

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
  • Printed Message: Warns of slips, trips and falls
  • Bright Color: Warns of presence of leaks, drips or spills even in low light

Specifications

Color Black, Red on Yellow
Capacity (Metric) 81 L
Material Polypropylene
Type Absorbent Pad
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 18.9 in.
Length (Metric) 48 cm
Width (English) 14.96 in.
Width (Metric) 38 cm
Alternate Part Number CH100-E
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.