missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents BrightSorb High Visibility Safety Heavyweight Absorbent Pad

Product Code. p-200113501
Click to view available options
Capacity (Metric):
81 L
155 L
263 L
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 30533356

Brand: Brady™ 309070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MAXX means greater performance without the extra weight. Specially developed chemical sorbent, yellow color-coded for safety and easy separation of hazardous waste, heavy weight pad. Does not react with aggressive fluids.

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
  • Printed Message: Warns of slips, trips and falls
  • Bright Color: Warns of presence of leaks, drips or spills even in low light

Specifications

Color Black, Red on Yellow
Capacity (Metric) 81 L
Material Polypropylene
Type Absorbent Pad
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 18.9 in.
Length (Metric) 48 cm
Width (English) 14.96 in.
Width (Metric) 38 cm
Alternate Part Number CH100-E
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.