missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Drain Cover Bags Small

Product Code. 30533392 Shop All Brady Products
Change view
Click to view available options
Length (Metric):
80 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
30533392 80 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533392 Supplier Brady™ Supplier No. 813923
  • ONLINE EXCLUSIVE PRICE
    91.50 €
    85.67 € / Pack of 1
    Save 5.83 €
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BAG-PU-S is a carry bag made from polyurethane that protects the PU50 and PU60 drain seal during storage and facilitates transport.

  • A chemical sorbent soc, filled with chemical meltblown polypropylene that absorbs chemicals and hazardous liquids.
  • Strong wicking action: absorption begins instantly after contact with the spilled liquid, no need to turn the soc.
  • Flexible design.

Specifications

Color Yellow
Type Drain Cover Bags
For Use With (Equipment) To Protect the PU50 and PU60 Drain Seals During Storage and to Facilitate Transport.
Model Spill Containment
Length (English) 31.496 in.
Length (Metric) 80 cm
Diameter (English) 7.874 in.
Diameter (Metric) 20.00 cm
Alternate Part Number BAG-PU-S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.