Learn More
Brady™ SPC™ Absorbents Oil Only Kit
Product Code. 30533387 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
48.872 gal.
Unit Size:
Each
Product Code. Capacity (English) Unit Size
30533387 48.872 gal. Each
1 options
Product Code. 30533387 Supplier Brady™ Supplier No. 813829
  • ONLINE EXCLUSIVE PRICE
    574.10 €
    535.50 € / Pack of 1
    Save 38.60 €
    6% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SA-AFO is a combination of a 2-ply, white, heavy weight, bonded and perforated oil only sorbent roll that absorbs oil and petroleum based liquids and a nitrile rubber anti-fatigue base mat. Special design provides buoyancy for basic fatigue relief. Silicone free allowing mat to be used in vehicle painting facilities.
SA-AFO is a combination of a 2-ply, white, heavy weight, bonded and perforated oil only sorbent roll (76 cm x 46 m, perfed every 76 cm) that absorbs oil and petroleum based liquids and a nitrile rubber anti-fatigue base mat (82 x 152 cm).
  • Special design provides buoyancy for basic fatigue relief.
  • Silicone free allowing mat to be used in vehicle painting facilities.
  • High quality nitrile rubber makes the anti-fatique mat oil resistant.
  • Designed to keeps workers safe while protecting the floor from oils and other industrial liquids.
Specifications
Color Gray
Capacity (English) 48.872 gal.
Capacity (Metric) 185 L
Type Absorbent Kit
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Process, Warehouse and Logistics
Includes One 76 cm x 46 m Perforated Roll and One Antifatigue Base Mat 82 x 152 cm
Model Oil Only Sorbent
Alternate Part Number SA-AFO