missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Oil Only Spill Kit, 12 Barrel Compliant

Product Code. 30533331 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
595 gal.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
30533331 595 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533331 Supplier Brady™ Supplier No. 813905
  • ONLINE EXCLUSIVE PRICE
    5778.10 €
    5410.64 € / Pack of 1
    Save 367.46 €
    6% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SL-12 BARR-C is 12 barrel spill kit that comes in two polyethylene spill containers. Specially designed conform the OPA regulations for fast spill response in case of oil spills on water.

SL-7 BARR-C is 7 barrel spill kit that comes in a polyethylene spill container.

  • Specially designed conform the OPA regulations for fast spill response in case of oil spills on water.
  • Contents: 750 pads (41 x 51 cm), 12 socs, (7.6 x122 cm), 8 pillows (43 x 48 cm), 12 booms (13 cm x 3 m), 1 sweep (48 cm x 30.5 m), 2 pair of goggles, 5 disposable jump suits, 5 pair of gloves, 2 non-spark scoops, 10 yellow disposal bags, 1 non-sparking pump, 2 x 20 L buckets and 2 anti-static shovels.

Specifications

Color Orange
Capacity (English) 595 gal.
Type Spill Kit
For Use With (Equipment) Emergency Spill Response
Includes 950 Pads (41 x 51 cm), 48 Socs, (7.6 x 122 cm), 8 Pillows (43 x 48 cm), 20 Booms (13 cm x 3 m), 4 Sweeps (48 x 30.5 m), 2 Pair of Goggles, 5 Disposable Jump Suits, 5 Pair of Gloves, 2 Non-spark Scoops, 10 Yellow Disposal Bags, 1 Non-sparking Pump, 2 x 20 L Buckets and 2 Anti-static Shovels.
Model Oil Only Sorbent
Alternate Part Number SL-12 BARR-C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.