missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Spill Containment Berm, QuickBerm Rigid Lock

Product Code. 30533386
Change view
Click to view available options
Capacity (Metric) Sump:
662 L
890 L
2820 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sump unitSize
30533386 662 L Each
30533325 890 L Each
30533399 2820 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30533386 Supplier Brady™ Supplier No. 198531
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness.

  • The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.

Specifications

Capacity (English) Sump 174.882 gal.
Capacity (Metric) Sump 662 L
Color Black
Material PVC-coated Fabric
Type Containment Berm
For Use With (Application) Beverage Processing, Chemical, Food Processing, Government, Healthcare, Manufacturing, Maritime, Mass transit, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Spill Containment
Length (English) 6.667 ft.
Length (Metric) 2.03 m
Width (English) 4.665 ft.
Width (Metric) 1.42 m
Height (English) 12.008 in.
Height (Metric) 30.50 cm
Alternate Part Number SB-QB-RL21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.