missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Spill Containment Berm, QuickBerm Rigid Lock

Product Code. 30533399 Shop All Brady Products
Change view
Click to view available options
Capacity (Metric) Sump:
662 L
890 L
2820 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sump Unit Size
30533399 2820 L Each
30533386 662 L Each
30533325 890 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30533399 Supplier Brady™ Supplier No. 198533
  • ONLINE EXCLUSIVE PRICE
    2875.60 €
    2689.36 € / Pack of 1
    Save 186.24 €
    6% Off
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness.

  • The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.

Specifications

Capacity (English) Sump 744.965 gal.
Capacity (Metric) Sump 2820 L
Color Black
Material PVC-coated Fabric
Type Containment Berm
For Use With (Application) Beverage Processing, Chemical, Food Processing, Government, Healthcare, Manufacturing, Maritime, Mass transit, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Spill Containment
Length (English) 10.827 ft.
Length (Metric) 3.30 m
Width (English) 10.827 ft.
Width (Metric) 3.30 m
Height (English) 12.008 in.
Height (Metric) 30.50 cm
Alternate Part Number SB-QB-RL33
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.