Learn More
Brady™ SPC™ Absorbents Spill Kit Container - Extra Large
Product Code. 30533363 Shop All Brady Products
Change view
Click to view available options
Size:
X-Large
Unit Size:
1 set
Product Code. Size Unit Size
30533363 X-Large 1 set
1 options
Product Code. 30533363 Supplier Brady™ Supplier No. 813907
  • ONLINE EXCLUSIVE PRICE
    731.00 €
    683.94 € / 1 sets
    Save 47.06 €
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SC-XLBIN is an extra large container made from chemical and water resistant polyethylene. Ideal for creating your own custom emergency response kits. Large enough to hold plenty of sorbents as well as any tools or minor equipment.
SC-XLBIN is an extra large container made from chemical and water resistant polyethylene.
  • Ideal for creating your own custom emergency response kits.
  • Large enough to hold plenty of sorbents as well as any tools or minor equipment.
Specifications
Color Orange
Material Polyethylene
Type Spill Kit Container
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Process, Warehouse and Logistics
Size X-Large
Weight (Metric) 32.5 kg
Alternate Part Number SC-XLBIN