missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Universal Plus Chemical Roll

Product Code. p-200113504
Click to view available options
Length (Metric):
23 m
46 m
Width (Metric):
38 cm
76 cm
Unit Size:
1 roll
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 30533351

Brand: Brady™ 813844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A 2-ply, green, medium weight, bonded and double perforated chemical sorbent roll that absorbs chemicals and hazardous liquids. One sided spunbound gives extra strength as well as less lint. Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.

A 2-ply, green, medium weight, bonded and double perforated chemical sorbent rolls that absorbs chemicals and hazardous liquids.

  • One sided spunbound gives extra strength as well as less lint.
  • Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.
  • Green color alerts workers that the product is being used for hazardous liquids.
  • Green color also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs.
  • Perforated down the middle and cross-wise to permit the use of precisely the right amount of material to reduce waste.
  • Perfect for cleaning up chemical spills or for use in laboratories.

Specifications

Color Green
Capacity (English) 40.947 gal.
Capacity (Metric) 155 L
Type Absorbent Roll
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 150.919 ft.
Length (Metric) 46 m
Width (English) 14.96 in.
Width (Metric) 38 cm
Alternate Part Number UN15-2DP-E
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.