missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Universal Plus Chemically Inert Chemical Roll

Product Code. 30533375 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
3.17 gal.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
30533375 3.17 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533375 Supplier Brady™ Supplier No. 834177
  • ONLINE EXCLUSIVE PRICE
    31.20 €
    29.21 € / Pack of 1
    Save 1.99 €
    6% Off
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One sided spunbond gives extra strength as well as lower lint. Chemically inert, product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid. Green color alerts workers that the product is being used for hazardous liquids.

One sided spunbond gives extra strength as well as lower lint.

  • Chemically inert, product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid.
  • Green color alerts workers that the product is being used for hazardous liquids.

Specifications

Color Green
Capacity (English) 3.17 gal.
Capacity (Metric) 12.00 L
Type Absorbent Roll
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 49.21 ft.
Length (Metric) 15 m
Width (English) 7.48 in.
Width (Metric) 19 cm
Alternate Part Number UN295-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.