missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Universal Plus Laboratory Chemical Roll

Product Code. 30533388 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
10.303 gal.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
30533388 10.303 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533388 Supplier Brady™ Supplier No. 322009
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A 2-ply, green, medium weight, bonded and double perforated chemical sorbent roll that absorbs chemicals and hazardous liquids.

  • One sided spunbound gives extra strength as well as less lint
  • Does not degrade or cause a dangerous chemical reaction with the absorbed liquid
  • Green color alerts workers that the product is being used for hazardous liquids
  • Green color also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Perforated down the middle and cross-wise to permit the use of precisely the right amount of material to reduce waste
  • Perfect for cleaning up chemical spills or for use in laboratories

Specifications

Color Green
Capacity (English) 10.303 gal.
Capacity (Metric) 39 L
Type Absorbent Roll
Model Chemical Sorbent
Length (English) 75.459 ft.
Length (Metric) 23 m
Width (English) 14.57 in.
Width (Metric) 37 cm
Alternate Part Number UN295-DPS-E-KADDIE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.