missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ MRO200-E Absorbents MRO Plus Perforated Absorbent Roll

Product Code. 30533382
Change view
Click to view available options
Capacity (English):
40.154 gal.
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) unitSize
30533382 40.154 gal. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533382 Supplier Brady™ Supplier No. 138588
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

MRO200-E is a 2-ply, grey, light weight, bonded and perforated universal sorbent pad that absorbs industrial liquids including oils, water, solvents, coolants, etc. One sided spunbound gives extra strength as well as less lint.

MRO200-E is a 2-ply, grey, light weight, bonded and perforated universal sorbent pad that absorbs industrial liquids including oils, water, solvents, coolants, etc.

  • One sided spunbound gives extra strength as well as less lint.
  • Perforated to help minimize waste and lower disposal costs.
  • Ideal sorbent for dealing with leaks and drips in the workplace.
  • Also suited for general industrial maintenance and spill clean up.

Specifications

Color Gray
Capacity (English) 40.154 gal.
Capacity (Metric) 152 L
Type Absorbent Roll
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Warehouse and Logistics
Length (English) 19.685 in.
Length (Metric) 50 cm
Width (English) 15.748 in.
Width (Metric) 40 cm
Alternate Part Number MRO200-E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.