missing translation for 'onlineSavingsMsg'
Learn More

SPD304, MedChemExpress

Product Code. 16481149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481149 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481149 Supplier Medchem Express Supplier No. HY111255
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SPD304 is a selective TNF-α inhibitor, which promotes dissociation of TNF trimers and therefore blocks the interaction of TNF and its receptor. SPD304 has an IC50 of 22 μM for inhibiting in vitro TNF receptor 1 (TNFR1) binding to TNF-α.

SPD304 is a selective TNF-α inhibitor, which promotes dissociation of TNF trimers and therefore blocks the interaction of TNF and its receptor. SPD304 has an IC50 of 22 μM for inhibiting in vitro TNF receptor 1 (TNFR1) binding to TNF-α.

Specifications

Chemical Name or Material SPD304
CAS 869998-49-2
Color White
Purity Grade Notes Research
Molecular Formula C32H32F3N3O2
Quantity 1 mg
Solubility Information DMSO : 25 mg/mL (45.65 mM; Need ultrasonic) ∣H2O : 20 mg/mL (36.52 mM; Need ultrasonic)
SMILES O=C1C(CN(C)CCN(C)CC2=CN(C3=CC=CC(C(F)(F)F)=C3)C4=C2C=CC=C4)=COC5=CC(C)=C(C)C=C15
Molecular Weight (g/mol) 547.61
Formula Weight 547.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.