missing translation for 'onlineSavingsMsg'
Learn More

SPDP, MedChemExpress

Product Code. 16494256
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494256 100 mg 100mg
16472848 10 mg 10mg
16471516 500 mg 500mg
16440906 200 mg 200mg
16439506 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494256 Supplier Medchem Express Supplier No. HY100216
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).

SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).

Specifications

Chemical Name or Material SPDP
CAS 68181-17-9
Color White
Purity Grade Notes Research
Molecular Formula C12H12N2O4S2
Quantity 100 mg
Synonym SPDP Crosslinker
Solubility Information DMSO : 155 mg/mL (496.22 mM; Need ultrasonic)
SMILES O=C(ON1C(CCC1=O)=O)CCSSC2=NC=CC=C2
Molecular Weight (g/mol) 312.36
Formula Weight 312.36
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.