Learn More
Invitrogen™ Custom DNA Oligos Synthesis Services
Custom Primer
Brand: Invitrogen™ 10629186
.15 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00500kg
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
Normal or Reversed-phase Chromatography, HPLC or PAGE | |
Custom Primer, 25nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification | |
Design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modification |
Store at room temperature. | |
Custom Primer | |
25 nmol |