missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Special Optical Glass 6030-OG Macro Cell Cuvettes

Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Special Optical Glass 6030-OG Macro Cell Cuvettes are manufactured from SCHOTT DESAG™, a colourless highly transparent crown glass in the form of drawn panes with fire-polished surfaces.

Brand:  Hellma™ 6030-40-10

Additional Details : Weight : 0.02700kg

 View more versions of this product

Product Code. 10015110

  • 122.00 € / Each

Estimated Shipment: 15-06-2023
to see stock.
Add to basket

Description

Description

  • Wavelength of 360 to 2500nm
  • For photometry and colorimetry measurements
Specifications

Specifications

Macro Absorption Cell
Special Optical Glass
40 mm
45mm
For photometry and colorimetry measurements
14,000 μL
9.5mm
360 to 2500nm
12.5mm
Rectangular
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers