Learn More
Bochem™ Special Retort Clamp
Product Code. 10106650
Change view
Click to view available options
Size (Metric) Grip:
0 to 70 mm
Unit Size:
Each
Product Code. Size (Metric) Grip Unit Size
10106650 0 to 70 mm Each
1 options
Product Code. 10106650 Supplier Bochem™ Supplier No. 5562
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Offers flexibility of use. BOCHEM™ Retort Special Retort Clamp is made of aluminum and PVC.
  • 3 prongs
  • Aluminum/PVC
Specifications
Quantity 1 Ea.
Length (Metric) Arm 140 mm
Size (Metric) Grip 0 to 70 mm