missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SpeciMAX™ Dx Stabilized Saliva Collection Kit

Product Code. 17478332
Click to view available options
Quantity:
100 Kits
Unit Size:
100 sets
This item is not returnable. View return policy

Product Code. 17478332

Brand: Thermo Scientific™ A51023

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intended for the collection and preservation of saliva specimens in the presence of healthcare professionals as an aid in nucleic acid-based Polymerase Chain Reaction (PCR) for In Vitro Diagnostic purposes.

  • Intended for the collection and preservation of saliva specimens in the presence of healthcare professionals
  • An aid in nucleic acid-based polymerase chain reaction (PCR) for in vitro diagnostic purpose
  • The individual single-use peel-pouch kit comes with an easy-to-use spill-resistant funnel, an automation-friendly saliva collection tube, and a screw cap to secure the sample after collection
  • The SpeciMAX Dx saliva collection tube is pre-filled with 1 mL of stabilization agent for virus inactivation
  • Integrates easily into automatic processing and analysis workflows that are part of high-throughput diagnostic testing
  • Refrigeration or freezing stabilized saliva samples is not recommended as precipitation of the stabilization agent has downstream effects

Features of the SpeciMAX Dx Stabilized Saliva Collection Kit include:

  • Inactivation of common respiratory viruses
  • Stabilization solution that preserves gDNA, RNA, and viral nucleic acid in saliva samples
  • Nonhazardous stabilization solution that is compatible with all disinfection protocols—no reaction with bleach (no guanidinium or ethanol)
  • Barcoded, standardized tubes and caps that fit automated workflows
  • Instrumental in high-throughput diagnostic testing

Sample storage

  • Refrigeration or freezing the stabilized saliva samples is not recommended as precipitation of the stabilization agent has downstream effects
  • The SpeciMAX stabilization agent preserves nucleic acid in saliva samples at ambient temperature for the following lengths of time:
    • gDNA: 6 months
    • RNA: 30 days
    • Viral nucleic acid: 30 days

Easy to use

  • Each SpeciMAX Dx Stabilized Saliva Collection Kit comes ready to use in individually packaged easy-peel blister pouches
  • To use the kit, the saliva donor follows four easy steps:
    • Remove and save the cap, then attach the funnel securely to the collection tube.
    • Fill saliva to the 2.0 line (not including bubbles)
    • Unscrew and discard the funnel (do not reuse)
    • Secure the cap on the tube, then mix by inverting the tube five times
  • For best results, saliva donors should be instructed to refrain from eating, drinking, using tobacco products, or chewing gum for at least 30 minutes prior to sample collection
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stabilized Saliva Collection Kit
Certifications/Compliance CE, IVD
Content And Storage • Virus inactivating and nucleic stabilization solution (1 mL)
• Pre-filled saliva collection tube, cap, and funnel

Store at 15&°C to 30°C.
For Use With (Application) For In Vitro Diagnostic Use
Includes Virus inactivating and nucleic stabilization solution (1 mL) pre-filled saliva collection tube, cap, and funnel
Quantity 100 Kits
Sterility Non-sterile
Storage Requirements Store at 15°C to 30°C
Volume (Metric) 6 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.