Learn More
Kartell™ Faeces Container
Easy means of collecting and transporting faeces samples. Kartell™ Faeces Container includes a spoon to assist in handling the sample.
Brand: Kartell™ 0259500
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Particularly designed for collection and delivery of faeces
- Made of transparent PS with stopper of PP
- Incorporates a small spoon to facilitate sample handling
- Sterile
- CE Marked and IVD Certified