missing translation for 'onlineSavingsMsg'
Learn More

Spectrum Labs™ Spectra/Por™ 3 Membrane Disc

Product Code. 12757526
Change view
Click to view available options
MWCO:
3.5 kDa
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
12757526 3.5 kDa Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12757526 Supplier Spectrum Labs™ Supplier No. 132486
  • ONLINE EXCLUSIVE PRICE
    286.00 €
    252.18 € / Pack of 50
    Save 33.82 €
    11% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems

For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant

Specifications

Product Type Membrane Disc
MWCO 3.5 kDa
Membrane Regenerated Cellulose
Diameter (Metric) 47 mm
For Use With (Application) For Higher Permeability
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.