missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 5 Pre-treated Dialysis Tubing

Product Code. 11485859
Change view
Click to view available options
MWCO:
12 to 14 kDa
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
11485859 12 to 14 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485859 Supplier Spectrum™ Labs Supplier No. 132754
  • ONLINE EXCLUSIVE PRICE
    427.00 €
    324.00 € / Pack of 1
    Save 103.00 €
    24% Off
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO

Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5.Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.
  • Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
  • Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
  • Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide For molecular separation, protein binding studies, electrolyte removal and desalting.

Specifications

Product Type Dialysis Tubing
MWCO 12 to 14 kDa
Diameter (Metric) 48 mm
Length (Metric) 15 m
Capacity (Metric) 18 mL/cm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.