missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Por™ Cellulose Regenerated Tubing Membrane
Click to view available options
MWCO:
12 to 14 kDa
15 kDa
25 kDa
50 kDa
8 kDa
Length (Metric):
10 m
15 m
5 m
Capacity (Metric):
0.32 mL/cm
0.45 mL/cm
1.1 mL/cm
46 mL/cm
5.1 mL/cm
6.4 mL/cm
Unit Size:
Each
Description
Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5. Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.
- Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
- Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
- Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide
Specifications
Specifications
| Product Type | Regenerated Tubing Membrane |
| MWCO | 8 kDa |
| Material | Cellulose |
| Diameter (Metric) | 7.5 mm |
| Length (Metric) | 10 m |
| Capacity (Metric) | 0.45 mL/cm |
| Color | Transparent |
| Temperature Range (Metric) | 60°C |
| Quantity | 1 Each |
| For Use With (Application) | For molecular separation, protein binding studies, electrolyte removal and desalting |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido