Learn More
Spectrum™ Labs Spectra/Por™ Membrane Kit
Product Code. 15370682
Click to view available options
Product Type:
Spectra/Por Membrane Kit
Unit Size:
1 set
Product Code. 15370682
Brand: Spectrum™ Labs 132 000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Spectra/Por Membrane Kit
Kit Contents Biotech grade regenerate Cellulose (rc) dialysis tubing in 3 different mwco, 7 spectra/por closures (3 standard, 3 weighted, magnetic-weighted), one spectra/por dialysis reservoir (1800ml) with stir bar
Quantity 1 Kit
For Use With (Application) Perfect solution for dialysis applications