Learn More
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer Accessories
Product Code. 11833784 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cell Holder
Cuvette Rack
Replacement Sample Compartment
SPECTRONIC Standards 2 Kit
Unit Size:
1 set
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11833784 Cuvette Rack Pack of 6
15287116 Cell Holder Each
15661794 Replacement Sample Compartment Each
15390187 SPECTRONIC Standards 2 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11833784 Supplier Thermo Scientific™ Supplier No. 222265600
  • ONLINE EXCLUSIVE PRICE
    69.65 €
    68.75 € / Pack of 6
    Save 0.90 €
    1% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
The SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer ideal for routine applications. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.
Rugged, Practical Design
  • With no internal parts, there's nothing to wear out
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from acidic or oxidizing solutions
  • Navigation pad and on-screen menus are second nature to any modern user
  • Compact footprint saves space in your lab
  • Cuvette racks on both sides of the sample compartment help you keep track of samples
  • No computer or serial cable required; Just plug it in and you're ready to measure
The Perfect Tool for Routine Spectroscopic Measurements
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Standard sample compartment accommodates both 10mm square cuvettes and test tubes up to 25mm (1 inch) diameter
  • Room light immune optical system lets you measure tall test tubes with the lid open
  • Analyzer mode allows you to program standard methods
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
  • Available Thermo Scientific VISIONlite5 software, for your computer, controls the instrument and provides data manipulation, storage and report printing options
Not ready to part with your old Thermo Scientific Spec 20 or GENESYS 20
  • The SPECTRONIC 200 will emulate the interface and controls on your old instrument to integrate seamlessly into your laboratory
VISIONlite5 software — Volume pricing begins at eight copies and is available on request.
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette Rack