Learn More
Shimadzu™ Spectrophotometer Biospec-Mini (DNA/RNA/Protein Analyser) Uv-Vis
Product Code. 12903806
Encompass
Change view
Click to view available options
Type:
Spectrophotometer Biospec-Mini
Unit Size:
Each
Product Code. Type Unit Size
12903806 Spectrophotometer Biospec-Mini Each
1 options
Product Code. 12903806 Supplier Shimadzu™ Supplier No. 2410625138
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
With on Std software and LCD monitor 220-240V 50/60Hz measuring range 190-1100nm bandwidth 5nm Shimadzu
Specifications
Type Spectrophotometer Biospec-Mini