Learn More
Bolle Safety Spectrum Goggle
Product Code. 31810813 Shop All Bollé Safety Products
Change view
Click to view available options
Frame Type:
Indirect Vented
Indirect Vented with Foam
Sealed
Sealed with Foam
Lens Tint:
Clear
Smoke
Model:
SPECTN10W
SPECTN11W
SPECTN12W
SPECTN13W
SPECTN14W
SPECTN15W
SPECTN20W
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Type Lens Tint Model Unit Size
31810813 Indirect Vented Smoke SPECTN20W Each
31810811 Sealed Clear SPECTN15W Each
31810799 Indirect Vented Clear SPECTN14W Each
31810775 Sealed with Foam Clear SPECTN13W Each
31810781 Sealed Clear SPECTN12W Each
31810787 Indirect Vented with Foam Clear SPECTN11W Each
31810806 Indirect Vented Clear SPECTN10W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31810813 Supplier Bolle Safety Supplier No. SPECTN20W
  • ONLINE EXCLUSIVE PRICE
    17.42 €
    16.03 € / Pack of 1
    Save 1.39 €
    7% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
With its bi-material frame and adjustable strap, Spectrum offers superior protection and adaptability. Available in two sizes, Spectrum is a versatile product with different frame types and lens coatings to provide a solution for every company, regardless of their specific work environment. Its sporty design and optimized ergonomics also ensure greater acceptance for enhanced safety and productivity.
  • PC/TPR Co-injected frame for optimum comfort.
  • Multiple coatings and sizes available.
  • Soft gasket to fit most face structures.
  • Optimal panoramic vision.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Platinum
Lens Tint Smoke
Product Type Safety Goggles
Model SPECTN20W
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Frame Type Indirect Vented
Autoclavable Not Autoclavable