missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing

Product Code. 13523880
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
MWCO:
20 kDa
3.5 to 5 kDa
8 to 10 kDa
Unit Size:
5m
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) MWCO Unit Size
13523880 6.4 mm 3.5 to 5 kDa Each
15370782 6.4 mm 8 to 10 kDa 5m
15390782 10 mm 8 to 10 kDa 5m
13542040 10 mm 20 kDa Each
15310792 6.4 mm 20 kDa 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 13523880 Supplier Spectrum™ Supplier No. 133192
  • ONLINE EXCLUSIVE PRICE
    519.00 €
    339.00 € / Pack of 1
    Save 180.00 €
    34% Off
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE

  • Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
  • Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure, does not interfere with molecular activity and function
  • 3 MWCOs and 2 tubing flat-widths
  • Packaged dry with glycerin (soak 15 min. in water)
  • Use with pH 2 to 12 and temp 4 to 60°C

Specifications

Product Type Spectra/Por Dialysis Tubing
MWCO 3.5 to 5 kDa
Material Regenerated Cellulose
Diameter (Metric) 6.4 mm
Diameter (English) 0.25 in.
Length (English) 16.4 ft.
Length (Metric) 5 m
Formulation Packaged dry with glycerin (soak 15 min. in water)
Temperature Range (English) 39.2°F to 140°F
Temperature Range (Metric) 4°C to 60°C
Quantity Each
For Use With (Application) Frequent dialysis applications, desalting, purification, sample preparation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.