missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
385.00 € - 431.00 €
Specifications
Product Type | Spectra/Por Dialysis Tubing |
---|---|
Material | Regenerated Cellulose |
Length (English) | 16.4 ft. |
Length (Metric) | 5 m |
Formulation | Packaged dry with glycerin (soak 15 min. in water) |
Product Code | Brand | Diameter (English) | Diameter (Metric) | MWCO | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | MWCO | Price | Quantity & Availability | |||
13523880
|
Spectrum™
133192 |
0.25 in. | 6.4 mm | 3.5 to 5 kDa |
431.00 €
Each |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15370782
|
Spectrum™
133264 |
0.25 in. | 6.4 mm | 8 to 10 kDa |
431.00 €
5m |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15390782
|
Spectrum™
133270 |
0.39 in. | 10 mm | 8 to 10 kDa |
385.00 €
5m |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13542040
|
Spectrum™
133342 |
0.39 in. | 10 mm | 20 kDa |
431.00 €
Each |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15310792
|
Spectrum™
133336 |
0.25 in. | 6.4 mm | 20 kDa |
431.00 €
5m |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
- Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
- Rigidly controlled porosity for better selectivity and separations
- Ultrapure, does not interfere with molecular activity and function
- 3 MWCOs and 2 tubing flat-widths
- Packaged dry with glycerin (soak 15 min. in water)
- Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Spectra/Por Dialysis Tubing | |
16.4 ft. | |
Packaged dry with glycerin (soak 15 min. in water) | |
4°C to 60°C | |
Frequent dialysis applications, desalting, purification, sample preparation |
Regenerated Cellulose | |
5 m | |
39.2°F to 140°F | |
Each |