Learn More
Spectrum™ Standard RC Ready-to-use Dialysis Sacks Spectra/Por™
Product Code. 15340752
Change view
Click to view available options
MWCO:
12 to 14 kDa
Unit Size:
Pack of 10
Product Code. MWCO Unit Size
15340752 12 to 14 kDa Pack of 10
1 options
Product Code. 15340752 Supplier Spectrum™ Supplier No. 132666
  • ONLINE EXCLUSIVE PRICE
    388.00 €
    356.19 € / Pack of 10
    Save 31.81 €
    8% Off
Estimated Shipment: 01-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Ready-to-Use Dialysis Sacks
MWCO 12 to 14 kDa
Membrane Spectra/Por 1
Width (Metric) 50 mm
Capacity (Metric) 50 to 400 mL