missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Brand:  Spectrum™ 132675

Code : LB

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11455849

  • 655.00 € / Each
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Specifications

Spectra/Por Dialysis Tubing
Regenerated Cellulose
3 in.
15 m
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
6 to 8 kDa
76 mm
49.2 ft.
39.2°F to 249.8°F
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers