missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO

Código de producto. 11475849
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
6.4 mm
16 mm
45 mm
67 mm
76 mm
missing translation for 'unitSize':
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Diameter (Metric) missing translation for 'unitSize'
11475849 16 mm Each
11405859 76 mm Each
11495849 67 mm Each
11465849 6.4 mm Each
11485849 45 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11475849 missing translation for 'mfr' Spectrum™ missing translation for 'supplierNo' 132678
  • ONLINE EXCLUSIVE PRICE
    823.00 €
    432.00 € / Pack of 1
    Save 391.00 €
    47% Off
In Stock
Añadir a la cesta
Edge
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Especificaciones

Product Type Spectra/Por 2 Dialysis Tubing
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Format Tubing
Diameter (Metric) 16 mm
Diameter (English) 0.62 in.
Length (English) 49.2 ft.
Length (Metric) 15 m
Formulation Contain glycerin as a humectant
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.