missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO

Product Code. 11485849
Change view
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
45 mm
67 mm
76 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11485849 45 mm Each
11475849 16 mm Each
11405859 76 mm Each
11495849 67 mm Each
11465849 6.4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11485849 Supplier Spectrum™ Supplier No. 132680
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 2 Dialysis Tubing
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 45 mm
Diameter (English) 1.77 in.
Length (English) 49.2 ft.
Length (Metric) 15 m
Formulation Contain glycerin as a humectant
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.