missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, TRC

Product Code. 30137826
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137826 25 g 25g
30132761 5 g 5g
30003572 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137826 Supplier Toronto Research Chemicals Supplier No. TRCS68040025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7 H19 N3
Molecular Weight (g/mol) 145.25
Synonym 1,4-Butanediamine, N1-(3-aminopropyl)-;1,4-Butanediamine, N-(3-aminopropyl)- (8CI,9CI), Spermidine (6CI), 1,5,10-Triazadecane, 1,8-Diamino-4-azaoctane, 4-Azaoctane-1,8-diamine, N-(3-Aminopropyl)-1,4-butanediamine, N-(3-Aminopropyl)-1,4-diaminobutane, N-(3-Aminopropyl)-4-aminobutylamine, N-(4-Aminobutyl)-1,3-diaminopropane, Spermidin
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine
SMILES NCCCCNCCCN

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2
Formula Weight 145.1579
Percent Purity >95%
Chemical Name or Material Spermidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.