missing translation for 'onlineSavingsMsg'
Learn More

Spermidine trihydrochloride, 99+%

Product Code. 15484169
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484169 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15484169 Supplier Thermo Scientific Alfa Aesar Supplier No. J61595.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 334-50-9 | C7H22Cl3N3 | 254.62 g/mol

Binds and precipitates DNA; may be used for purification of DNA binding proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 334-50-9
Melting Point 250°C (decomposition)
Color White
Molecular Formula C7H22Cl3N3
Linear Formula NH2(CH2)3NH(CH2)4NH2·3HCl
MDL Number MFCD00012918
Quantity 100 mg
Beilstein 3552356
Merck Index 14,8742
Synonym spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride
Solubility Information Soluble in water
InChI Key LCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride
Molecular Weight (g/mol) 254.62
PubChem CID 9539
Formula Weight 254.63
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Spermidine trihydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.