missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermidine Trihydrochloride, Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
25g
5g
Beschreibung
Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms
Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins
Spermidine Trihydrochloride forms stable compounds with nucleic acids. It is used in biochemical research.
Spezifikation
Spezifikation
| Melting Point | 248°C |
| Color | White |
| pH | 3 to 6 |
| Assay Percent Range | ≥99 % |
| Packaging | Amber Glass |
| Quantity | 1 g |
| Merck Index | 15, 8869 |
| Formula Weight | 254.63 |
| Percent Purity | 99% min. |
| Physical Form | Solid |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Spermidine trihydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur