missing translation for 'onlineSavingsMsg'
Learn More

Spermine, 97%

Product Code. 10795781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795781 1 g Glass bottle 1g
10488480 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795781 Supplier Thermo Scientific Acros Supplier No. 132750010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 71-44-3 | C10H26N4 | 202.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name N,N'-bis(3-aminopropyl)butane-1,4-diamine
SMILES NCCCNCCCCNCCCN

Specifications

Melting Point 26.0°C to 30.0°C
Density 0.9370g/mL
Boiling Point 130.0°C (0.1 mmHg)
Flash Point 110°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2N(CH2)3NH(CH2)4NH(CH2)3NH2
Quantity 1 g
Beilstein 04, II, 704
Merck Index 15, 8870
Specific Gravity 0.937
Formula Weight 202.34
Percent Purity 97%
Chemical Name or Material Spermine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Spermine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.