missing translation for 'onlineSavingsMsg'
Learn More

Spermine, free base, ≥97%, MP Biomedicals™

Product Code. 11444381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444381 1 g 1g
11454381 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444381 Supplier MP Biomedicals Supplier No. 0215207001
  • ONLINE EXCLUSIVE PRICE
    88.60 €
    84.00 € / 1g
    Save 4.60 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
SMILES NCCCNCCCCNCCCN

Specifications

Color White
Quantity 1 g
Formula Weight 202.3
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Spermine
compliance-icons
Product Identifier
  • Spermine Free Base
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Spermine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.