missing translation for 'onlineSavingsMsg'
Learn More

Spermine, free base, ≥97%, MP Biomedicals™

Product Code. 11454381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454381 5 g 5g
11444381 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454381 Supplier MP Biomedicals Supplier No. 0215207005
  • ONLINE EXCLUSIVE PRICE
    355.00 €
    336.00 € / 5g
    Save 19.00 €
    5% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
SMILES NCCCNCCCCNCCCN

Specifications

Color White
Quantity 5 g
Formula Weight 202.3
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Spermine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.