missing translation for 'onlineSavingsMsg'
Learn More

Spermine Tetrahydrochloride, MP Biomedicals

Product Code. 11450525 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450525 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11450525 Supplier MP Biomedicals Supplier No. 0210047410
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Used to precipitate DNA from low salt aqueous buffers
  • Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors and red blood cells of chicken

Specifications

CAS 306-67-2
Melting Point 312°C to 314°C (literature)
Color White
Molecular Formula C10H30Cl4N4
Linear Formula C10H26N4 · 4HCl
MDL Number MFCD00012914
Quantity 10 g
Synonym spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride
Solubility Information Soluble in water (100mg/mL - clear, colorless solution).
InChI Key XLDKUDAXZWHPFH-UHFFFAOYSA-N
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
IUPAC Name tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride
Molecular Weight (g/mol) 348.18
PubChem CID 9384
Formula Weight 348.2
Percent Purity ≥96%
Physical Form Powder
Chemical Name or Material Spermine Tetrahydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Spermine Tetrahydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contain: Spermine Tetrahydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.