Learn More
SPEX CertiPrep™ OdorEroder
Product Code. 15286106 Shop All SPEX Certiprep Products
Change view
Click to view available options
Volume (Metric):
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15286106 100 g 100g
15296106 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15286106 Supplier SPEX CertiPrep™ Supplier No. ODER100G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Effectively neutralizes a wide range of chemical odors in the lab
  • Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
  • As odors get trapped, the beads start turning brown
Specifications
For Use With (Application) Effectively neutralizes a wide range of chemical odors in the lab
Volume (Metric) 100 g